Assay Details
| Gene Symbol: Rab2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155967.1 | - | 2687 | 101 | ||
| AK171898.1 | - | 2722 | 101 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab2b| Gene Symbol: Rab2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155967.1 | - | 2687 | 101 | ||
| AK171898.1 | - | 2722 | 101 | |||
Set Membership: |
|