Assay Details
| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024439.3 | NP_077759.3 | 6 | 119 | ||
| GenBank mRNA | AF335543.1 | - | 6 | 119 | ||
| AI836862.1 | - | 1 | 231 | 119 | ||
| AK005204.1 | - | 6 | 119 | |||
| AK131659.1 | - | 1 | 119 | |||
| BC011091.1 | - | 6 | 3983625 | 119 | ||
| BC096566.1 | - | 3 | 119 | |||
More Information
Set Membership: |
|

Vimp
Genomic Map