Assay Details
| Gene Symbol: Slc25a51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009949.3 | NP_001009949.3 | 3 | 3876 | 108 | |
| GenBank mRNA | AK146788.1 | - | 3 | 3838 | 108 | |
| AK156168.1 | - | 5 | 3907 | 108 | ||
| AK156342.1 | - | 3 | 3834 | 108 | ||
| BC089000.1 | - | 2 | 3771 | 6418170 | 108 | |
More Information
Set Membership: |
|

Slc25a51
Genomic Map