Assay Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009981.2 | NP_001009981.1 | 11 | 2028 | 78 | |
| NM_001282108.1 | NP_001269037.1 | 10 | 1986 | 78 | ||
| NM_001282109.1 | NP_001269038.1 | 11 | 2036 | 78 | ||
| NM_029736.2 | NP_084012.1 | 12 | 2112 | 78 | ||
| NR_104094.1 | 10 | 1955 | 78 | |||
| GenBank mRNA | AK051650.1 | - | 8 | 1797 | 78 | |
| AK079901.1 | - | 10 | 1963 | 78 | ||
| AK152752.1 | - | 12 | 2071 | 78 | ||
| AK152849.1 | - | 12 | 2071 | 78 | ||
| AK165983.1 | - | 11 | 1974 | 78 | ||
More Information
Set Membership: |
|

Slc10a7
Genomic Map