Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145857.2 | NP_665856.2 | 2 | 192 | 87 | |
| GenBank mRNA | AF520774.1 | - | 2 | 237 | 87 | |
| AK089390.1 | - | 2 | 156 | 87 | ||
| AK089843.1 | - | 2 | 156 | 87 | ||
| AY160220.1 | - | 1 | 121 | 87 | ||
| AY160221.1 | - | 2 | 193 | 87 | ||
| BC044774.2 | - | 2 | 153 | 87 | ||
More Information
Set Membership: |
|

Nod2
Genomic Map