Assay Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010421.4 | NP_034551.2 | 1 | 143 | 94 | |
| GenBank mRNA | AK075895.1 | - | 1 | 119 | 94 | |
| AK075911.1 | - | 1 | 144 | 94 | ||
| AK080777.1 | - | 1 | 116 | 94 | ||
| AK144168.1 | - | 1 | 271 | 94 | ||
| AK150444.1 | - | 1 | 114 | 94 | ||
| AK151960.1 | - | 1 | 118 | 94 | ||
| AK159091.1 | - | 1 | 135 | 94 | ||
| AK159814.1 | - | 1 | 129 | 94 | ||
| AK159890.1 | - | 1 | 116 | 94 | ||
| AK160112.1 | - | 1 | 103 | 94 | ||
| AK167013.1 | - | 1 | 129 | 94 | ||
| AK168186.1 | - | 1 | 143 | 94 | ||
| AK170903.1 | - | 1 | 129 | 94 | ||
| BC010755.1 | - | 1 | 112 | 4225233 | 94 | |
| U07631.1 | - | 1 | 135 | 94 | ||
| X64331.1 | - | 1 | 86 | 94 | ||
More Information
Set Membership: |
|

Hexa
Genomic Map