Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016797.4 | NP_058077.2 | 10 | 2025 | 93 | |
| GenBank mRNA | AK082927.1 | - | 10 | 1665 | 93 | |
| AK142742.1 | - | 1672 | 93 | |||
| AK158923.1 | - | 10 | 2026 | 93 | ||
More Information
Set Membership: |
|

Stx7
Genomic Map