Assay Details
| Gene Symbol: Rfx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033536.1 | NP_001028708.1 | 9 | 3820 | 123 | |
| GenBank mRNA | AK037109.1 | - | 1 | 306 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Rfx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033536.1 | NP_001028708.1 | 9 | 3820 | 123 | |
| GenBank mRNA | AK037109.1 | - | 1 | 306 | 123 | |
Set Membership: |
|