Assay Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009034.4 | NP_033060.3 | 1 | 110 | 77 | |
| GenBank mRNA | AK003312.1 | - | 1 | 95 | 77 | |
| BC125551.1 | - | 1 | 79 | 40130066 | 77 | |
| BC125553.1 | - | 1 | 79 | 40130068 | 77 | |
| X74154.1 | - | 1 | 94 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003312.1, BC125551.1, BC125553.1, NM_009034.4, X74154.1 transcripts.
Set Membership: |
|

Rbp2
Genomic Map