Assay Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007542.4 | NP_031568.2 | 2 | 262 | 93 | |
| GenBank mRNA | AK086920.1 | - | 2 | 259 | 93 | |
| AK135496.1 | - | 2 | 263 | 93 | ||
| AK155973.1 | - | 2 | 263 | 93 | ||
| AK164867.1 | - | 2 | 263 | 93 | ||
| AK171870.1 | - | 2 | 263 | 93 | ||
| BC005452.1 | - | 2 | 256 | 3594469 | 93 | |
| BC019502.1 | - | 2 | 265 | 4237268 | 93 | |
| BC052857.1 | - | 2 | 247 | 30061850 | 93 | |
| BC057185.1 | - | 2 | 274 | 93 | ||
| L20276.1 | - | 2 | 262 | 93 | ||
| X53928.1 | - | 357 | 93 | |||
| Y11758.1 | - | 1 | 93 | 93 | ||
More Information
Set Membership: |
|

Bgn
Genomic Map