Assay Details
| Gene Symbol: Dynll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168471.1 | NP_001161943.1 | 3 | 696 | 88 | |
| NM_001168472.1 | NP_001161944.1 | 3 | 869 | 88 | ||
| NM_026556.4 | NP_080832.1 | 3 | 902 | 88 | ||
| GenBank mRNA | AK028319.1 | - | 3 | 667 | 88 | |
| AK049645.1 | - | 3 | 876 | 88 | ||
| AK078434.1 | - | 3 | 875 | 88 | ||
| AK160114.1 | - | 3 | 872 | 88 | ||
| AK169212.1 | - | 3 | 875 | 88 | ||
| BC011289.1 | - | 3 | 693 | 3981362 | 88 | |
| BC040822.1 | - | 3 | 864 | 4459858 | 88 | |
| BC058797.1 | - | 3 | 512 | 88 | ||
More Information
Set Membership: |
|

Dynll2
Genomic Map