Assay Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028149.1 | NP_082425.1 | 15 | 2589 | 84 | |
| GenBank mRNA | AK144786.1 | - | 15 | 2563 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl20| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028149.1 | NP_082425.1 | 15 | 2589 | 84 | |
| GenBank mRNA | AK144786.1 | - | 15 | 2563 | 84 | |
Set Membership: |
|