Assay Details
| Gene Symbol: Gnb2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008143.3 | NP_032169.1 | 1 | 105 | 77 | |
| GenBank mRNA | AK002493.1 | - | 1 | 94 | 77 | |
| AK017772.1 | - | 1 | 106 | 77 | ||
| AK051226.1 | - | 1 | 106 | 77 | ||
| AK159797.1 | - | 1 | 106 | 77 | ||
| AK160528.1 | - | 1 | 76 | 77 | ||
| AK160739.1 | - | 1 | 103 | 77 | ||
| AK161234.1 | - | 1 | 106 | 77 | ||
| AK166417.1 | - | 1 | 94 | 77 | ||
| AK166786.1 | - | 1 | 108 | 77 | ||
| AK166800.1 | - | 1 | 109 | 77 | ||
| AK166868.1 | - | 1 | 134 | 77 | ||
| AK166945.1 | - | 1 | 106 | 77 | ||
| AK168086.1 | - | 1 | 106 | 77 | ||
| AK168096.1 | - | 1 | 106 | 77 | ||
| AK168196.1 | - | 1 | 106 | 77 | ||
| AK168349.1 | - | 1 | 137 | 77 | ||
| AK169612.1 | - | 1 | 106 | 77 | ||
| BC020322.1 | - | 1 | 82 | 77 | ||
| BC046760.1 | - | 1 | 90 | 5693613 | 77 | |
| D29802.1 | - | 1 | 99 | 77 | ||
| X75313.1 | - | 1 | 105 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002493.1, AK017772.1, AK051226.1, AK159797.1, AK160528.1, AK160739.1, AK161234.1, AK166417.1, AK166786.1, AK166800.1, AK166945.1, AK168086.1, AK168096.1, AK168196.1, AK168349.1, BC046760.1, D29802.1, NM_008143.3, X75313.1 transcripts.
Set Membership: |
|

Gnb2l1
Genomic Map