Assay Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109657.3 | NP_001103127.1 | 1 | 106 | 68 | |
| GenBank mRNA | AK031455.1 | - | 1 | 107 | 68 | |
| BB620887.1 | - | 1 | 107 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109657.3 transcripts.
Set Membership: |
|

Gas7
Genomic Map