Assay Details
| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027107.1 | NP_081383.1 | 4 | 519 | 63 | |
| GenBank mRNA | AK009156.1 | - | 4 | 486 | 63 | |
| AK146257.1 | - | 4 | 509 | 63 | ||
| BC027639.1 | - | 4 | 464 | 1069092 | 63 | |
| BQ030877.1 | - | 1 | 63 | |||
More Information
Set Membership: |
|

Ten1
Genomic Map