Assay Details
| Gene Symbol: Wars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164314.1 | NP_001157786.1 | 10 | 1910 | 93 | |
| NM_001164488.1 | NP_001157960.1 | 10 | 1843 | 93 | ||
| GenBank mRNA | AK004541.1 | - | 10 | 1844 | 93 | |
| AK150090.1 | - | 11 | 1874 | 93 | ||
| AK150267.1 | - | 11 | 1957 | 93 | ||
| AK152966.1 | - | 10 | 1874 | 93 | ||
| AK153337.1 | - | 10 | 1881 | 93 | ||
| BC046232.1 | - | 10 | 1849 | 6813406 | 93 | |
More Information
Set Membership: |
|

Wars
Genomic Map