Assay Details
| Gene Symbol: Glrx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028419.2 | NP_082695.1 | 2 | 751 | 139 | |
| GenBank mRNA | AK009158.1 | - | 2 | 756 | 139 | |
| AK013761.1 | - | 2 | 752 | 139 | ||
| AK050883.1 | - | 751 | 139 | |||
| AK148847.1 | - | 1 | 1037 | 139 | ||
| BC050937.1 | - | 2 | 771 | 139 | ||
| BC058371.1 | - | 2 | 771 | 6819353 | 139 | |
More Information
Set Membership: |
|

Glrx5
Genomic Map