Assay Details
| Gene Symbol: Gm17750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI851080.1 | - | 1 | 90 | ||
| AK082072.1 | - | 2 | 371 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17750| Gene Symbol: Gm17750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI851080.1 | - | 1 | 90 | ||
| AK082072.1 | - | 2 | 371 | 90 | ||
Set Membership: |
|