Assay Details
| Gene Symbol: Nespas | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002846.2 | 1 | 294 | 84 | |||
| GenBank mRNA | AF319945.1 | - | LIT1669 Nespas |
1 | 247 | 84 | |
| AK153813.1 | - | 1 | 295 | 84 | |||
More Information
Set Membership: |
|

Nespas
Genomic Map