Assay Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301340.1 | NP_001288269.1 | 16 | 1925 | 85 | |
| NM_001301341.1 | NP_001288270.1 | 18 | 2052 | 85 | ||
| NM_001301343.1 | NP_001288272.1 | 16 | 1911 | 85 | ||
| NM_001301344.1 | NP_001288273.1 | 15 | 1853 | 85 | ||
| NM_001301345.1 | NP_001288274.1 | 15 | 1793 | 85 | ||
| NM_025279.3 | NP_079555.1 | 16 | 1865 | 85 | ||
| GenBank mRNA | AK011428.1 | - | 16 | 1870 | 85 | |
| AK051313.1 | - | 15 | 1787 | 85 | ||
| AK078777.1 | - | 18 | 2053 | 85 | ||
| AK088462.1 | - | 15 | 1779 | 85 | ||
| AK128891.1 | - | 16 | 1861 | 85 | ||
| AK135519.1 | - | 16 | 1839 | 85 | ||
| AK144245.1 | - | 9135 | 85 | |||
| AK150848.1 | - | 16 | 1905 | 85 | ||
| AK151689.1 | - | 15 | 1806 | 85 | ||
| AK152927.1 | - | 15 | 1807 | 85 | ||
| AK160615.1 | - | 16 | 1895 | 85 | ||
| AK160887.1 | - | 16 | 1866 | 85 | ||
| AK166656.1 | - | 16 | 1930 | 85 | ||
| AK166927.1 | - | 16 | 1884 | 85 | ||
| AK167600.1 | - | 16 | 1856 | 85 | ||
| AK168918.1 | - | 12 | 1766 | 85 | ||
| AK169064.1 | - | 16 | 1915 | 85 | ||
| AK186860.1 | - | 1 | 352 | 85 | ||
| AK192668.1 | - | 1 | 351 | 85 | ||
| AK196607.1 | - | 1 | 322 | 85 | ||
| AK206192.1 | - | 1 | 264 | 85 | ||
| BC006694.1 | - | 16 | 1892 | 3583037 | 85 | |
| BC089328.1 | - | 16 | 1849 | 6402614 | 85 | |
More Information
Set Membership: |
|

Hnrnpk
Genomic Map