Assay Details
| Gene Symbol: Ccdc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK161058.1 | - | 3 | 2150 | 87 | |
| AK162990.1 | - | 3 | 2141 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc55| Gene Symbol: Ccdc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK161058.1 | - | 3 | 2150 | 87 | |
| AK162990.1 | - | 3 | 2141 | 87 | ||
Set Membership: |
|