Assay Details
| Gene Symbol: B3gnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169114.1 | NP_001162585.1 | 1 - 2 | 119 | 78 | |
| GenBank mRNA | BC009075.1 | - | 1 - 2 | 119 | 2654354 | 78 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC009075.1, NM_001169114.1 transcripts.
Set Membership: |
|

B3gnt2
Genomic Map