Assay Details
| Gene Symbol: Prok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170419.1 | NP_001163890.1 | 1 - 2 | 165 | 88 | |
| GenBank mRNA | BB614931.1 | - | 1 - 2 | 157 | 88 | |
More Information
Set Membership: |
|

Prok2
Genomic Map