Assay Details
| Gene Symbol: Mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171187.1 | NP_001164658.1 | 1 - 2 | 661 | 79 | |
| GenBank mRNA | AK019046.1 | - | 1 - 2 | 187 | 79 | |
More Information
Set Membership: |
|

Mal
Genomic Map