Assay Details
| Gene Symbol: Eid2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177427.1 | NP_001170898.1 | 1 | 327 | 98 | |
| GenBank mRNA | AK044722.1 | - | 1 | 296 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eid2b| Gene Symbol: Eid2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177427.1 | NP_001170898.1 | 1 | 327 | 98 | |
| GenBank mRNA | AK044722.1 | - | 1 | 296 | 98 | |
Set Membership: |
|