Assay Details
| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177703.1 | NP_001171174.1 | 1 - 2 | 207 | 73 | |
| GenBank mRNA | AK007645.1 | - | 1 - 2 | 207 | 73 | |
| CX239940.1 | - | 1 - 2 | 119 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007645.1, NM_001177703.1 transcripts.
Set Membership: |
|

Gal3st1
Genomic Map