Assay Details
| Gene Symbol: Sstr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191008.1 | NP_001177937.1 | 1 - 2 | 168 | 68 | |
| GenBank mRNA | AK133767.1 | - | 1 - 2 | 96 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133767.1, NM_001191008.1 transcripts.
Set Membership: |
|

Sstr5
Genomic Map