Assay Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111051.1 | NP_001104521.1 | 7 - 8 | 936 | 73 | |
| NM_001111052.1 | NP_001104522.1 | 7 - 8 | 936 | 73 | ||
| NM_001195538.1 | NP_001182467.1 | 11 - 12 | 1797 | 73 | ||
| NM_001195539.1 | NP_001182468.1 | 7 - 8 | 984 | 73 | ||
| NM_019978.3 | NP_064362.1 | 11 - 12 | 1845 | 73 | ||
| GenBank mRNA | AB093234.1 | - | 11 - 12 | 1754 | 73 | |
| AF155819.1 | - | 11 - 12 | 1743 | 73 | ||
| AF155821.1 | - | 7 - 8 | 795 | 73 | ||
| AK032424.1 | - | 7 - 8 | 826 | 73 | ||
| AK047716.1 | - | 7 - 8 | 723 | 73 | ||
| AK081200.1 | - | 7 - 8 | 779 | 73 | ||
| AY968047.1 | - | 7 - 8 | 684 | 73 | ||
| AY968048.1 | - | 7 - 8 | 687 | 73 | ||
| BC133685.1 | - | 11 - 12 | 1599 | 40110683 | 73 | |
More Information
Set Membership: |
|

Dclk1
Genomic Map