Assay Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111051.1 | NP_001104521.1 | 5 - 6 | 674 | 94 | |
| NM_001111052.1 | NP_001104522.1 | 5 - 6 | 674 | 94 | ||
| NM_001195538.1 | NP_001182467.1 | 9 - 10 | 1535 | 94 | ||
| NM_001195539.1 | NP_001182468.1 | 5 - 6 | 722 | 94 | ||
| NM_019978.3 | NP_064362.1 | 9 - 10 | 1583 | 94 | ||
| GenBank mRNA | AB093234.1 | - | 9 - 10 | 1492 | 94 | |
| AF155819.1 | - | 9 - 10 | 1481 | 94 | ||
| AF155820.1 | - | 9 - 10 | 1474 | 94 | ||
| AF155821.1 | - | 5 - 6 | 533 | 94 | ||
| AK032424.1 | - | 5 - 6 | 564 | 94 | ||
| AK047716.1 | - | 5 - 6 | 461 | 94 | ||
| AK081200.1 | - | 5 - 6 | 517 | 94 | ||
| AY968047.1 | - | 5 - 6 | 422 | 94 | ||
| AY968048.1 | - | 5 - 6 | 425 | 94 | ||
| BC133685.1 | - | 9 - 10 | 1337 | 40110683 | 94 | |
More Information
Set Membership: |
|

Dclk1
Genomic Map