Assay Details
| Gene Symbol: Smlr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195596.1 | NP_001182525.1 | 1 - 2 | 313 | 82 | |
| GenBank mRNA | AK008073.1 | - | 1 - 2 | 311 | 82 | |
More Information
Set Membership: |
|

Smlr1
Genomic Map