Assay Details
| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198933.1 | NP_001185862.1 | 10 - 11 | 1368 | 98 | |
| NM_008615.2 | NP_032641.2 | 10 - 11 | 1290 | 98 | ||
| GenBank mRNA | AK004980.1 | - | 10 - 11 | 1156 | 98 | |
| AK006387.1 | - | 4 - 5 | 411 | 98 | ||
| AK077968.1 | - | 10 - 11 | 1158 | 98 | ||
| AK163403.1 | - | 10 - 11 | 1368 | 98 | ||
| BC003287.1 | - | 10 - 11 | 1141 | 3582779 | 98 | |
| BC011081.1 | - | 10 - 11 | 1188 | 3978265 | 98 | |
| BC080660.1 | - | 10 - 11 | 1146 | 4240732 | 98 | |
| BC094028.1 | - | 10 - 11 | 1145 | 4974808 | 98 | |
| J02652.1 | - | 10 - 11 | 1191 | 98 | ||
| M26756.1 | - | 10 - 11 | 1133 | 98 | ||
| M29546.1 | - | 402 | 98 | |||
More Information
Set Membership: |
|

Me1
Genomic Map