Assay Details
| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198933.1 | NP_001185862.1 | 3 - 4 | 599 | 74 | |
| NM_008615.2 | NP_032641.2 | 3 - 4 | 521 | 74 | ||
| GenBank mRNA | AK004980.1 | - | 3 - 4 | 387 | 74 | |
| AK077968.1 | - | 3 - 4 | 389 | 74 | ||
| AK163278.1 | - | 3 - 4 | 599 | 74 | ||
| AK163403.1 | - | 3 - 4 | 599 | 74 | ||
| BC003287.1 | - | 3 - 4 | 378 | 3582779 | 74 | |
| BC011081.1 | - | 3 - 4 | 419 | 3978265 | 74 | |
| BC080660.1 | - | 3 - 4 | 377 | 4240732 | 74 | |
| BC094028.1 | - | 3 - 4 | 376 | 4974808 | 74 | |
| J02652.1 | - | 3 - 4 | 422 | 74 | ||
| M26756.1 | - | 3 - 4 | 364 | 74 | ||
More Information
Set Membership: |
|

Me1
Genomic Map