Assay Details
| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198933.1 | NP_001185862.1 | 6 - 7 | 943 | 106 | |
| NM_008615.2 | NP_032641.2 | 6 - 7 | 865 | 106 | ||
| GenBank mRNA | AK004980.1 | - | 6 - 7 | 731 | 106 | |
| AK077968.1 | - | 6 - 7 | 733 | 106 | ||
| AK163278.1 | - | 6 - 7 | 943 | 106 | ||
| AK163403.1 | - | 6 - 7 | 943 | 106 | ||
| BC003287.1 | - | 6 - 7 | 722 | 3582779 | 106 | |
| BC011081.1 | - | 6 - 7 | 763 | 3978265 | 106 | |
| BC080660.1 | - | 6 - 7 | 721 | 4240732 | 106 | |
| BC094028.1 | - | 6 - 7 | 720 | 4974808 | 106 | |
| J02652.1 | - | 6 - 7 | 766 | 106 | ||
| M26756.1 | - | 6 - 7 | 708 | 106 | ||
More Information
Set Membership: |
|

Me1
Genomic Map