Assay Details
| Gene Symbol: Serpina3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199940.1 | NP_001186869.1 | 2 - 3 | 226 | 119 | |
| GenBank mRNA | BY136799.1 | - | 2 - 3 | 227 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001199940.1 transcripts.
Set Membership: |
|

Serpina3i
Genomic Map