Assay Details
| Gene Symbol: B230216G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242345.1 | NP_001229274.1 | 4 - 5 | 215 | 82 | |
| GenBank mRNA | AK043509.1 | - | 2 - 3 | 128 | 82 | |
| AK045625.1 | - | 1 - 2 | 309 | 82 | ||
More Information
Set Membership: |
|

B230216G23Rik
Genomic Map