Assay Details
| Gene Symbol: A630095N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243090.1 | NP_001230019.1 | 2 - 3 | 250 | 70 | |
| NM_001243091.1 | NP_001230020.1 | 2 - 3 | 163 | 70 | ||
| GenBank mRNA | AK080399.1 | - | 2 - 3 | 164 | 70 | |
More Information
Set Membership: |
|

A630095N17Rik
Genomic Map