Assay Details
| Gene Symbol: Ddx39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252457.1 | NP_001239386.1 | 10 - 11 | 1422 | 62 | |
| NM_019693.3 | NP_062667.1 | 10 - 11 | 1510 | 62 | ||
| GenBank mRNA | AA231260.1 | - | 1 - 2 | 80 | 62 | |
| AK051034.1 | - | 10 - 11 | 1443 | 62 | ||
| AK088867.1 | - | 10 - 11 | 1428 | 62 | ||
| AK202224.1 | - | 71 | 62 | |||
| AK216397.1 | - | 1 - 2 | 71 | 62 | ||
| BC011067.1 | - | 10 - 11 | 1418 | 3498520 | 62 | |
| BC024859.1 | - | 9 - 10 | 2646 | 5359825 | 62 | |
More Information
Set Membership: |
|

Ddx39b
Genomic Map