Assay Details
| Gene Symbol: Cant1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025617.2 | NP_001020788.1 | 3 - 4 | 933 | 69 | |
| NM_001025618.2 | NP_001020789.1 | 3 - 4 | 891 | 69 | ||
| NM_001267591.1 | NP_001254520.1 | 2 - 3 | 734 | 69 | ||
| NM_029502.3 | NP_083778.2 | 3 - 4 | 1085 | 69 | ||
| GenBank mRNA | AK049879.1 | - | 3 - 4 | 827 | 69 | |
| AK081118.1 | - | 3 - 4 | 990 | 69 | ||
| AK085059.1 | - | 2 - 3 | 2181 | 69 | ||
| AK166384.1 | - | 2 - 3 | 2095 | 69 | ||
| BC020003.1 | - | 2 - 3 | 733 | 4208801 | 69 | |
More Information
Set Membership: |
|

Cant1
Genomic Map