Assay Details
| Gene Symbol: Cldn34c4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281537.1 | NP_001268466.1 | 1 - 2 | 131 | 79 | |
| GenBank mRNA | AK015117.1 | - | 1 - 2 | 132 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281537.1 transcripts.
Set Membership: |
|

Cldn34c4
Genomic Map