Assay Details
| Gene Symbol: Tomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282088.1 | NP_001269017.1 | 1 - 2 | 67 | 98 | |
| GenBank mRNA | AK158024.1 | - | 68 | 98 | ||
| EU627091.1 | - | 1 - 2 | 67 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the EU627091.1, NM_001282088.1 transcripts.
Set Membership: |
|

Tomt
Genomic Map