Assay Details
| Gene Symbol: Cd63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042580.1 | NP_001036045.1 | 7 - 8 | 791 | 60 | |
| NM_001282966.1 | NP_001269895.1 | 7 - 8 | 835 | 60 | ||
| NM_007653.3 | NP_031679.1 | 7 - 8 | 750 | 60 | ||
| GenBank mRNA | AF159427.1 | - | 7 - 8 | 671 | 60 | |
| AK028220.1 | - | 7 - 8 | 688 | 60 | ||
| AK146618.1 | - | 7 - 8 | 738 | 60 | ||
| AK150647.1 | - | 7 - 8 | 745 | 60 | ||
| AK151702.1 | - | 7 - 8 | 716 | 60 | ||
| AK151963.1 | - | 7 - 8 | 690 | 60 | ||
| AK152072.1 | - | 7 - 8 | 712 | 60 | ||
| AK152372.1 | - | 7 - 8 | 716 | 60 | ||
| AK159875.1 | - | 7 - 8 | 713 | 60 | ||
| AK163785.1 | - | 7 - 8 | 712 | 60 | ||
| AK166543.1 | - | 7 - 8 | 711 | 60 | ||
| AK166766.1 | - | 7 - 8 | 710 | 60 | ||
| AK170344.1 | - | 7 - 8 | 745 | 60 | ||
| AK211715.1 | - | 2 - 3 | 173 | 60 | ||
| BC008108.1 | - | 7 - 8 | 697 | 3157993 | 60 | |
| BC012212.1 | - | 7 - 8 | 697 | 3157993 | 60 | |
| BC083161.1 | - | 7 - 8 | 682 | 6530532 | 60 | |
| BP769648.1 | - | 1 - 2 | 169 | 60 | ||
| CT010179.1 | - | 6 - 7 | 646 | 60 | ||
| D16432.1 | - | 7 - 8 | 700 | 60 | ||
More Information
Set Membership: |
|

Cd63
Genomic Map