Assay Details
| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284369.1 | NP_001271298.1 | 3 - 4 | 338 | 117 | |
| NM_001284371.1 | NP_001271300.1 | 3 - 4 | 338 | 117 | ||
| NM_001284373.1 | NP_001271302.1 | 3 - 4 | 338 | 117 | ||
| NM_001284374.1 | NP_001271303.1 | 3 - 4 | 338 | 117 | ||
| GenBank mRNA | AK087424.1 | - | 3 - 4 | 326 | 117 | |
| AK139515.1 | - | 3 - 4 | 253 | 117 | ||
| AK139844.1 | - | 3 - 4 | 251 | 117 | ||
| AK140855.1 | - | 3 - 4 | 294 | 117 | ||
| AK145887.1 | - | 3 - 4 | 338 | 117 | ||
| AK146839.1 | - | 3 - 4 | 250 | 117 | ||
| BC082596.1 | - | 3 - 4 | 284 | 30649207 | 117 | |
| BC085137.1 | - | 3 - 4 | 281 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087424.1, AK139844.1, AK140855.1, AK145887.1, BC082596.1, NM_001284369.1, NM_001284371.1, NM_001284373.1, NM_001284374.1 transcripts.
Set Membership: |
|

Atf2
Genomic Map