Assay Details
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286383.1 | NP_001273312.1 | 2 - 3 | 393 | 136 | |
| GenBank mRNA | AY046597.1 | - | 2 - 3 | 353 | 136 | |
More Information
Set Membership: |
|

Cklf
Genomic Map