Assay Details
| Gene Symbol: Sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286538.1 | NP_001273467.1 | 3 - 4 | 554 | 68 | |
| NM_001286605.1 | NP_001273534.1 | 3 - 4 | 507 | 68 | ||
| GenBank mRNA | AY390765.1 | - | 3 - 4 | 447 | 68 | |
More Information
Set Membership: |
|

Sigmar1
Genomic Map