Assay Details
| Gene Symbol: Zfp180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045486.2 | NP_001038951.1 | 2 - 3 | 422 | 91 | |
| NM_001289640.1 | NP_001276569.1 | 2 - 3 | 422 | 91 | ||
| NM_001289641.1 | NP_001276570.1 | 2 - 3 | 422 | 91 | ||
| GenBank mRNA | AK141922.1 | - | 2 - 3 | 375 | 91 | |
| AK144892.1 | - | 2 - 3 | 423 | 91 | ||
| AK145004.1 | - | 2 - 3 | 387 | 91 | ||
| AK154182.1 | - | 2 - 3 | 379 | 91 | ||
| BC063741.1 | - | 2 - 3 | 365 | 6504558 | 91 | |
| BC066115.1 | - | 2 - 3 | 384 | 30619470 | 91 | |
| BC085472.1 | - | 2 - 3 | 395 | 6417473 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC063741.1, NM_001289640.1, NM_001289641.1 transcripts.
Set Membership: |
|

Zfp180
Genomic Map