Assay Details
| Gene Symbol: Prrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164275.2 | NP_001157747.1 | 2 - 3 | 151 | 76 | |
| NM_001289683.1 | NP_001276612.1 | 3 - 4 | 247 | 76 | ||
| NM_027322.2 | NP_081598.1 | 2 - 3 | 178 | 76 | ||
| GenBank mRNA | AK135895.1 | - | 3 - 4 | 242 | 76 | |
| AK161802.1 | - | 2 - 3 | 177 | 76 | ||
| BC032926.1 | - | 2 - 3 | 128 | 3375373 | 76 | |
| BE629723.1 | - | 2 - 3 | 128 | 76 | ||
| CJ052035.1 | - | 2 - 3 | 176 | 76 | ||
| CX242928.1 | - | 2 - 3 | 129 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC032926.1, NM_001164275.2, NM_001289683.1, NM_027322.2 transcripts.
Set Membership: |
|

Prrg1
Genomic Map