Assay Details
| Gene Symbol: Bcl2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289716.1 | NP_001276645.1 | 2 - 3 | 1003 | 75 | |
| NM_001289717.1 | NP_001276646.1 | 2 - 3 | 848 | 75 | ||
| NM_009743.5 | NP_033873.3 | 2 - 3 | 797 | 75 | ||
| GenBank mRNA | AK146461.1 | - | 2 - 3 | 953 | 75 | |
| AK150028.1 | - | 2 - 3 | 740 | 75 | ||
| AK172250.1 | - | 2 - 3 | 764 | 75 | ||
| BC089016.1 | - | 2 - 3 | 751 | 30606568 | 75 | |
| BC089017.1 | - | 1 - 2 | 588 | 30605282 | 75 | |
| L35049.1 | - | 2 - 3 | 801 | 75 | ||
| U10101.1 | - | 1 - 2 | 559 | 75 | ||
| U51278.1 | - | 1 - 2 | 661 | 75 | ||
| X83574.1 | - | 1 - 2 | 559 | 75 | ||
More Information
Set Membership: |
|

Bcl2l1
Genomic Map