Assay Details
| Gene Symbol: Ovol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289817.1 | NP_001276746.1 | 1 - 2 | 1394 | 114 | |
| GenBank mRNA | BF715622.1 | - | 2 - 3 | 261 | 114 | |
More Information
Set Membership: |
|

Ovol3
Genomic Map