Assay Details
| Gene Symbol: G6pc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289857.1 | NP_001276786.1 | 1 - 2 | 301 | 135 | |
| NM_021331.4 | NP_067306.1 | 1 - 2 | 301 | 135 | ||
| GenBank mRNA | AK148450.1 | - | 1 - 2 | 302 | 135 | |
| AK148465.1 | - | 1 - 2 | 253 | 135 | ||
| BC111905.1 | - | 1 - 2 | 276 | 135 | ||
| Z47787.1 | - | 1 - 2 | 279 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111905.1 transcripts.
Set Membership: |
|

G6pc2
Genomic Map