Assay Details
| Gene Symbol: Gm21992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290127.1 | NP_001277056.1 | 1 - 2 | 462 | 70 | |
| GenBank mRNA | EU287941.1 | - | 1 - 2 | 336 | 70 | |
More Information
Set Membership: |
|

Gm21992
Genomic Map